Assay Details
Target Gene Details
Entrez Gene ID: | 76795 |
Gene Name: | TBC1 domain family, member 9B |
Gene Aliases: |
- |
Location: |
Chr.11:50131057-50172785 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d9b | NM_001290759.1 | 12 | 2031 | NP_001277688.1 |
| NM_001290760.1 | 12 | 2031 | NP_001277689.1 | |
| NM_029745.2 | 12 | 2031 | NP_084021.2 | |
| AK035457.1 | 12 | 1996 | BAC29068.1 | |
| AK049999.1 | 12 | 1991 | BAC34024.2 | |
| AK088860.1 | 7 | 981 | ||
| AK147964.1 | 12 | 1996 | BAE28254.1 | |
| AK172999.1 | 13 | 1971 | BAD32277.1 | |
| AK178319.1 | 3 | 222 | ||
| AK179480.1 | 3 | 221 | ||
| AK184516.1 | 3 | 221 | ||
| AK187856.1 | ||||
| AK187922.1 | ||||
| AK189310.1 | ||||
| AK204235.1 | ||||
| BC048085.1 | 3 | 345 | AAH48085.1 | |
| BC062928.1 | 12 | 2018 | AAH62928.1 | |
| BC065080.1 | 12 | 2010 | AAH65080.1 |
More Information
Set Membership: |
|

Genomic Map