Assay Details
Target Gene Details
Entrez Gene ID: | 237930 |
Gene Name: | tubulin tyrosine ligase-like family, member 6 |
Gene Aliases: |
4932418K24Rik, D11Moh43e, D11Moh44e, t8130b59 |
Location: |
Chr.11:96133786-96165452 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll6 | NM_172799.4 | 13 | 1948 | NP_766387.2 |
| AK077033.1 | 12 | 2435 | BAC36577.1 | |
| AK145771.1 | 1 | 49 | BAE26641.1 | |
| AM690749.1 | 13 | 1948 | ||
| BC132203.1 | 12 | 2076 | ||
| BC138058.1 | 12 | 2076 | ||
| BC145130.1 | 12 | 2077 | ||
| BC145131.1 | 12 | 2076 |
More Information
Set Membership: |
|

Genomic Map