Assay Details
Target Gene Details
Entrez Gene ID: | 117160 |
Gene Name: | tweety homolog 2 (Drosophila) |
Gene Aliases: |
1110001A03Rik |
Location: |
Chr.11:114675468-114720984 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttyh2 | NM_053273.2 | 14 | 3144 | NP_444503.2 |
| AK003193.1 | 1 | 296 | ||
| AK152172.1 | 14 | 3096 | BAE31004.1 | |
| AK154835.1 | 14 | 3151 | ||
| AK155166.1 | 14 | 3145 | BAE33089.1 | |
| BC060740.1 | 14 | 3087 | AAH60740.1 |
More Information
Set Membership: |
|

Genomic Map