Assay Details
Target Gene Details
Entrez Gene ID: | 103733 |
Gene Name: | tubulin, gamma 1 |
Gene Aliases: |
1500010O08Rik, AI451582, AI503389, Tubg |
Location: |
Chr.11:101120131-101126419 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubg1 | NM_134024.2 | 11 | 1483 | NP_598785.1 |
| AB158480.1 | 11 | 1495 | BAD27264.1 | |
| AK154019.1 | 11 | 1451 | BAE32319.1 | |
| AK160125.1 | 11 | 1497 | ||
| BC006581.1 | 11 | 1483 | AAH06581.1 |
More Information
Set Membership: |
|

Genomic Map