Assay Details
Target Gene Details
Entrez Gene ID: | 237858 |
Gene Name: | tumor suppressor candidate 5 |
Gene Aliases: |
C130069F04Rik, DSPB1, Lost1 |
Location: |
Chr.11:76679873-76698664 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tusc5 | NM_177709.3 | 1 | 439 | NP_808377.1 |
| AK048522.1 | 1 | 439 | BAC33359.1 | |
| AK158395.1 | 1 | 402 | BAE34485.1 | |
| BC119801.1 | 1 | 235 | ||
| BC122875.1 | 1 | 235 |

Genomic Map