Assay Details
Target Gene Details
Entrez Gene ID: | 71888 |
Gene Name: | keratin 33A |
Gene Aliases: |
2310015J09Rik |
Location: |
Chr.11:100011199-100016212 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt33a | NM_027983.3 | 7 | 1338 | NP_082259.2 |
| AK009363.1 | 7 | 1339 | BAB26243.1 | |
| AK037251.1 | 7 | 1339 | BAC29777.1 | |
| BC029257.1 | 7 | 1321 | AAH29257.1 |

Genomic Map