Assay Details
Target Gene Details
Entrez Gene ID: | 406221 |
Gene Name: | keratin 40 |
Gene Aliases: |
Ka36 |
Location: |
Chr.11:99537485-99543203 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt40 | NM_001039666.1 | 1 | 372 | NP_001034755.1 |
| BC107359.1 | 1 | 372 | AAI07360.1 | |
| BC107360.2 | 1 | 370 | ||
| BC119521.2 | 1 | 372 | ||
| BC119522.2 | 1 | 372 |

Genomic Map