Assay Details
Target Gene Details
Entrez Gene ID: | 68239 |
Gene Name: | keratin 42 |
Gene Aliases: |
2410039E07Rik, K17n, Ka22, ecat6 |
Location: |
Chr.11:100262882-100272565 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt42 | NM_212483.2 | 8 | 1364 | NP_997648.2 |
| AK019115.1 | 5 | 881 | ||
| AK019133.1 | 5 | 826 | BAB31560.1 | |
| AY459292.1 | 8 | 1364 | AAR22526.1 | |
| BC112371.1 | 8 | 1356 | AAI12372.1 | |
| BC148204.1 | 8 | 1364 |

Genomic Map