Assay Details
Target Gene Details
Entrez Gene ID: | 18121 |
Gene Name: | noggin |
Gene Aliases: |
- |
Location: |
Chr.11:89300638-89302559 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nog | NM_008711.2 | 1 | 640 | NP_032737.1 |
| AK146488.1 | 1 | 641 | ||
| BC132527.1 | 1 | 166 | ||
| BC138148.1 | 1 | 166 |
More Information
Set Membership: |
|

Genomic Map