Assay Details
Target Gene Details
Entrez Gene ID: | 59040 |
Gene Name: | ras homolog gene family, member T1 |
Gene Aliases: |
2210403N23Rik, AA415293, AF244542, AI834919, Arht1, C430039G08Rik, Miro1 |
Location: |
Chr.11:80209005-80267907 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhot1 | NM_001163354.1 | 1 | 225 | NP_001156826.1 |
| NM_001163355.1 | 1 | 225 | NP_001156827.1 | |
| NM_021536.7 | 1 | 225 | NP_067511.4 | |
| AK019059.1 | 1 | 185 | BAB31529.2 | |
| AK021245.1 | 1 | 186 | ||
| AK028597.1 | 1 | 168 | BAC26025.1 | |
| AK032096.1 | 1 | 197 | ||
| AK041119.1 | 1 | 161 | BAC30828.1 | |
| AK043323.1 | 1 | 84 | ||
| AK053263.1 | 1 | 135 | ||
| AK054472.1 | 1 | 196 | BAC35792.1 | |
| AK078955.1 | 1 | 168 | BAC37478.1 | |
| AK138830.1 | 1 | 166 | BAE23792.1 | |
| AK141621.1 | 1 | 52 | BAE24771.1 | |
| AK147749.1 | 1 | 204 | BAE28113.1 | |
| AK147801.1 | 1 | 230 | BAE28148.1 | |
| AK167502.1 | 1 | 225 | BAE39578.1 | |
| AK169811.1 | 1 | 194 | ||
| BC046785.1 | 1 | 208 | AAH46785.1 | |
| BC058350.1 | 1 | 140 | AAH58350.1 |
More Information
Set Membership: |
|

Genomic Map