Assay Details
Target Gene Details
Entrez Gene ID: | 59040 |
Gene Name: | ras homolog gene family, member T1 |
Gene Aliases: |
2210403N23Rik, AA415293, AF244542, AI834919, Arht1, C430039G08Rik, Miro1 |
Location: |
Chr.11:80209005-80267907 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhot1 | NM_001163354.1 | 20 | 3865 | NP_001156826.1 |
| NM_001163355.1 | 20 | 3892 | NP_001156827.1 | |
| NM_021536.7 | 19 | 3769 | NP_067511.4 | |
| AK138830.1 | 19 | 3710 | BAE23792.1 |
More Information
Set Membership: |
|

Genomic Map