Assay Details
Target Gene Details
Entrez Gene ID: | 76795 |
Gene Name: | TBC1 domain family, member 9B |
Gene Aliases: |
- |
Location: |
Chr.11:50131057-50172785 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d9b | NM_001290759.1 | 22 | 4544 | NP_001277688.1 |
| NM_001290760.1 | 22 | 4547 | NP_001277689.1 | |
| NM_029745.2 | 21 | 4496 | NP_084021.2 | |
| AK031992.1 | 7 | 1966 | BAC27640.1 | |
| BC058596.1 | 9 | 2081 | AAH58596.1 |
More Information
Set Membership: |
|

Genomic Map