Assay Details
Target Gene Details
Entrez Gene ID: | 76795 |
Gene Name: | TBC1 domain family, member 9B |
Gene Aliases: |
- |
Location: |
Chr.11:50131057-50172785 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d9b | NM_001290759.1 | 5 | 742 | NP_001277688.1 |
| NM_001290760.1 | 5 | 742 | NP_001277689.1 | |
| NM_029745.2 | 5 | 742 | NP_084021.2 | |
| AK035457.1 | 5 | 707 | BAC29068.1 | |
| AK041820.1 | ||||
| AK049999.1 | 5 | 702 | BAC34024.2 | |
| AK147964.1 | 5 | 707 | BAE28254.1 | |
| AK166472.1 | 5 | 725 | ||
| AK172999.1 | 6 | 682 | BAD32277.1 | |
| AK181462.1 | 1 | 61 | ||
| AK182440.1 | 1 | 61 | ||
| AK182514.1 | 1 | 61 | ||
| AK198179.1 | 1 | 61 | ||
| AK201473.1 | 1 | 61 | ||
| AK211841.1 | 1 | 61 | ||
| AK212246.1 | 1 | 61 | ||
| BC062928.1 | 5 | 729 | AAH62928.1 | |
| BC065080.1 | 5 | 721 | AAH65080.1 |
More Information
Set Membership: |
|

Genomic Map