Assay Details
Target Gene Details
Entrez Gene ID: | 66330 |
Gene Name: | RIKEN cDNA 1700020L24 gene |
Gene Aliases: |
- |
Location: |
Chr.11:83437694-83441235 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700020L24Rik | NM_025492.3 | 3 | 653 | NP_079768.2 |
| AK005809.1 | 3 | 652 | BAB24249.1 | |
| AK006174.1 | 3 | 654 | BAC25136.1 | |
| BC152320.1 | 3 | 640 |

Genomic Map