Assay Details
Target Gene Details
Entrez Gene ID: | 66156 |
Gene Name: | anaphase promoting complex subunit 11 |
Gene Aliases: |
1110011I19Rik, R75218 |
Location: |
Chr.11:120598421-120608198 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anapc11 | NM_001038230.2 | 3 | 1929 | NP_001033319.1 |
| NM_025389.4 | 3 | 1774 | NP_079665.1 | |
| AK045408.1 | 3 | 1717 | BAC32348.1 |
More Information
Set Membership: |
|

Genomic Map