Assay Details
Target Gene Details
Entrez Gene ID: | 69664 |
Gene Name: | keratin associated protein 1-5 |
Gene Aliases: |
2310043L02Rik, AI592983 |
Location: |
Chr.11:99579977-99581016 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap1-5 | NM_027157.3 | 1 | 515 | NP_081433.2 |
| AK009782.1 | 1 | 517 | BAB26502.1 | |
| BC147095.1 | 1 | 490 | ||
| BC147096.1 | 1 | 490 |

Genomic Map