Assay Details
Target Gene Details
Entrez Gene ID: | 77914 |
Gene Name: | keratin associated protein 17-1 |
Gene Aliases: |
A030006P16Rik |
Location: |
Chr.11:99993232-99993994 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap17-1 | NM_001099774.2 | 1 | 378 | NP_001093244.1 |
| AK020698.1 | 1 | 379 |

Genomic Map