Assay Details
Target Gene Details
Entrez Gene ID: | 71453 |
Gene Name: | keratin associated protein 2-4 |
Gene Aliases: |
5530401P19Rik, KAP2.4, KRTAP2.4 |
Location: |
Chr.11:99614017-99614846 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap2-4 | NM_027800.1 | 1 | 561 | NP_082076.1 |
| AI892716.1 | 1 | 270 | ||
| AK017442.1 | 1 | 561 | BAB30746.1 |

Genomic Map