Assay Details
Target Gene Details
Entrez Gene ID: | 665891 |
Gene Name: | keratin associated protein 4-1 |
Gene Aliases: |
A030010K20Rik |
Location: |
Chr.11:99627229-99628239 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap4-1 | NM_001048196.1 | 1 | 842 | NP_001041661.1 |
| AK137752.1 | 1 | 843 | BAE23490.1 | |
| BC147087.1 | 1 | 813 | ||
| BC147088.1 | 1 | 813 |

Genomic Map