Assay Details
Target Gene Details
Entrez Gene ID: | 68673 |
Gene Name: | keratin associated protein 4-2 |
Gene Aliases: |
1110033F04Rik, AI604442 |
Location: |
Chr.11:99634114-99635084 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap4-2 | NM_026807.2 | 1 | 818 | NP_081083.2 |
| AK004055.1 | 1 | 819 | BAB23145.1 |

Genomic Map