Assay Details
Target Gene Details
Entrez Gene ID: | 68768 |
Gene Name: | keratin associated protein 4-6 |
Gene Aliases: |
1110054P19Rik |
Location: |
Chr.11:99665043-99665960 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap4-6 | NM_026834.2 | 1 | 765 | NP_081110.1 |
| AK004258.1 | 1 | 766 | BAE43167.1 |

Genomic Map