Assay Details
Target Gene Details
Entrez Gene ID: | 12295 |
Gene Name: | calcium channel, voltage-dependent, beta 1 subunit |
Gene Aliases: |
CAB1, Cchb1, Cchlb1 |
Location: |
Chr.11:98001508-98022975 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacnb1 | NM_001282977.1 | 14 | 1898 | NP_001269906.1 |
| NM_145121.3 | 14 | 1763 | NP_660099.2 | |
| AB685743.1 | 14 | 1882 | ||
| AY094173.1 | 14 | 1561 | AAM11474.1 |
More Information
Set Membership: |
|

Genomic Map