Assay Details
Target Gene Details
Entrez Gene ID: | 12323 |
Gene Name: | calcium/calmodulin-dependent protein kinase II, beta |
Gene Aliases: |
- |
Location: |
Chr.11:5969642-6065748 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camk2b | NM_001174053.1 | 20 | 2938 | NP_001167524.1 |
| NM_001174054.1 | 21 | 2890 | NP_001167525.1 | |
| NM_007595.5 | 21 | 2929 | NP_031621.3 | |
| AK046454.1 | 21 | 2930 | BAC32736.1 | |
| AK158794.1 | 16 | 2573 | BAE34670.1 | |
| AK163698.1 | 21 | 2746 | BAE37464.1 | |
| BC080273.1 | 20 | 2740 | AAH80273.1 | |
| X63615.1 | 21 | 2744 | CAA45160.1 |
More Information
Set Membership: |
|

Genomic Map