Assay Details
Target Gene Details
Entrez Gene ID: | 67579 |
Gene Name: | cytoplasmic polyadenylation element binding protein 4 |
Gene Aliases: |
Cpe-bp4, Cpeb-4, mCPEB-4 |
Location: |
Chr.11:31870884-31935635 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpeb4 | NM_001290676.1 | 9 | 3510 | NP_001277605.1 |
| NM_001290678.1 | 8 | 3459 | NP_001277607.1 | |
| NM_026252.4 | 10 | 3534 | NP_080528.2 | |
| AK015381.1 | 7 | 929 | BAB29821.1 | |
| AK015401.1 | 6 | 917 | BAB29832.1 | |
| AK021394.1 | 3 | 423 | ||
| AK173229.1 | 8 | 3468 | BAD32507.1 | |
| AY313775.1 | 10 | 2264 | AAQ20844.1 | |
| BC115430.1 | 8 | 2183 | ||
| BC115431.1 | 8 | 2183 | ||
| BC145863.1 | ||||
| BC145865.1 |
More Information
Set Membership: |
|

Genomic Map