Assay Details
Target Gene Details
Entrez Gene ID: | 75586 |
Gene Name: | keratin associated protein 9-3 |
Gene Aliases: |
2310040M23Rik |
Location: |
Chr.11:99597348-99598106 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap9-3 | NM_029351.2 | 1 | 636 | NP_083627.2 |
| AK009730.1 | 1 | 638 | BAB26467.1 | |
| AK131927.1 | 1 | 637 | BAE20877.1 |

Genomic Map