Assay Details
Target Gene Details
Entrez Gene ID: | 216049 |
Gene Name: | zinc finger protein 365 |
Gene Aliases: |
AI839779, AV340892, DBZ, Su48, Znf365, talanin |
Location: |
Chr.10:67886105-67912662 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp365 | NM_178679.2 | 1 | 51 | NP_848794.1 |
| AK122387.1 | BAC65669.1 | |||
| BC080272.1 | AAH80272.1 | |||
| CB519185.1 | 1 | 51 |

Genomic Map