Assay Details
Target Gene Details
Entrez Gene ID: | 50773 |
Gene Name: | 5',3'-nucleotidase, cytosolic |
Gene Aliases: |
Dnt, Dnt1, Umph-2, Umph2 |
Location: |
Chr.11:115490425-115491876 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nt5c | NM_015807.1 | 5 | 594 | NP_056622.1 |
| AF078840.1 | 5 | 594 | AAF36421.1 | |
| AK007418.1 | 5 | 633 | BAB25027.1 | |
| BC024368.1 | 5 | 597 | AAH24368.1 |

Genomic Map