Assay Details
Target Gene Details
Entrez Gene ID: | 52653 |
Gene Name: | NudC domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.11:40733661-40740046 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudcd2 | NM_001290697.1 | 1 | 401 | NP_001277626.1 |
| NM_026023.5 | 1 | 401 | NP_080299.4 | |
| AK004388.1 | 1 | 395 | BAB23283.1 | |
| AK005909.1 | 1 | 396 | BAB24313.1 | |
| AK010850.1 | 1 | 208 | BAB27222.1 | |
| AK012388.1 | 1 | 179 | BAB28205.1 | |
| AK031779.1 | 1 | 179 | BAC27545.1 | |
| AK133774.1 | 1 | 405 | ||
| AK218319.1 | ||||
| BC005646.1 | 1 | 162 | AAH05646.1 | |
| BE949482.1 | 1 | 121 | ||
| CJ083042.1 |
More Information
Set Membership: |
|

Genomic Map