Assay Details
Target Gene Details
Entrez Gene ID: | 209586 |
Gene Name: | NudC domain containing 3 |
Gene Aliases: |
AI427847, BC024322, mKIAA1068 |
Location: |
Chr.11:6105691-6200862 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudcd3 | NM_173748.4 | 2 | 724 | NP_776109.1 |
| AK030050.1 | 2 | 449 | ||
| AK036694.1 | 2 | 428 | ||
| AK046219.1 | 2 | 446 | BAC32642.1 | |
| AK049739.1 | 2 | 436 | BAC33899.1 | |
| AK050476.1 | 2 | 414 | BAC34277.1 | |
| AK159391.1 | 2 | 456 | BAE35045.1 | |
| AK159458.1 | 2 | 456 | BAE35100.1 | |
| AK169797.1 | 2 | 460 | BAE41373.1 | |
| AK170329.1 | 2 | 457 | BAE41723.1 | |
| AK171224.1 | 2 | 439 | BAE42326.1 | |
| BC024322.1 | 2 | 442 | AAH24322.3 | |
| BC037090.1 | 2 | 423 | AAH37090.1 | |
| BC057603.1 | 1 | 192 | AAH57603.1 | |
| CF738060.1 | ||||
| EU350933.1 | 2 | 415 |
More Information
Set Membership: |
|

Genomic Map