Assay Details
Target Gene Details
Entrez Gene ID: | 70325 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class W |
Gene Aliases: |
2610044A17Rik, Gwt1 |
Location: |
Chr.11:84876312-84880285 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigw | NM_001077636.1 | 2 | 1163 | NP_001071104.1 |
| NM_027388.2 | 2 | 1582 | NP_081664.2 | |
| AK081337.1 | 2 | 1583 | BAC38199.1 | |
| AK086537.1 | 2 | 1164 | BAC39687.1 | |
| BC144942.1 | 2 | 1516 |
More Information
Set Membership: |
|

Genomic Map