Assay Details
Target Gene Details
Entrez Gene ID: | 20556 |
Gene Name: | schlafen 2 |
Gene Aliases: |
Shlf2 |
Location: |
Chr.11:83065112-83070678 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slfn2 | NM_011408.1 | 2 | 702 | NP_035538.1 |
| AF099973.1 | 2 | 702 | AAC83826.1 | |
| BC138749.1 | 2 | 657 | ||
| BC138750.1 | 2 | 657 |

Genomic Map