Assay Details
Target Gene Details
Entrez Gene ID: | 216825 |
Gene Name: | ubiquitin specific peptidase 22 |
Gene Aliases: |
AI427806 |
Location: |
Chr.11:61151781-61175059 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp22 | NM_001004143.4 | 13 | 2568 | NP_001004143.2 |
| AK034492.1 | 13 | 2543 | ||
| AK084022.1 | 9 | 1726 | BAC39100.1 | |
| AK155071.1 | 13 | 2569 | BAE33026.1 | |
| AK160992.1 | 13 | 2477 | BAE36137.1 | |
| AK191821.1 | 1 | 203 | ||
| AK193842.1 | 1 | 203 | ||
| AK198335.1 | 1 | 203 | ||
| AK199010.1 | 1 | 203 | ||
| AK220368.1 | 13 | 2529 | BAD90248.1 | |
| BC058419.1 | 6 | 1387 | AAH58419.1 | |
| BC080737.1 | 13 | 2439 | AAH80737.1 |
More Information
Set Membership: |
|

Genomic Map