Assay Details
Target Gene Details
Entrez Gene ID: | 72046 |
Gene Name: | upregulator of cell proliferation |
Gene Aliases: |
2010005J08Rik, 9130001I21Rik, AW060359, Urg4, mKIAA1507 |
Location: |
Chr.11:5713417-5762436 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Urgcp | NM_001077661.1 | 6 | 1495 | NP_001071129.1 |
| NM_178623.3 | 6 | 1079 | NP_848738.2 | |
| AK033577.1 | 6 | 1053 | BAC28370.1 | |
| AK035580.1 | 6 | 1491 | BAC29111.1 | |
| AK041953.1 | 7 | 1744 | BAC31109.1 | |
| AK129378.1 | 5 | 1137 | BAC98188.1 | |
| AK166495.1 | 5 | 979 | BAE38807.1 | |
| BC066994.1 | 6 | 1376 | AAH66994.1 |

Genomic Map