Assay Details
Target Gene Details
Entrez Gene ID: | 11840 |
Gene Name: | ADP-ribosylation factor 1 |
Gene Aliases: |
- |
Location: |
Chr.11:59211411-59228267 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arf1 | NM_001130408.1 | 1 | 101 | NP_001123880.1 |
| NM_007476.3 | NP_031502.1 | |||
| AK077542.1 | ||||
| AK148458.1 | BAE28566.1 | |||
| AK148873.1 | BAE28685.1 | |||
| AK150092.1 | BAE29300.1 | |||
| AK150699.1 | BAE29778.1 | |||
| AK151223.1 | BAE30215.1 | |||
| AK151225.1 | BAE30217.1 | |||
| AK151696.1 | BAE30620.1 | |||
| AK151821.1 | BAE30718.1 | |||
| AK152015.1 | BAE30877.1 | |||
| AK152382.1 | BAE31170.1 | |||
| AK152479.1 | 1 | 115 | BAE31253.1 | |
| AK153132.1 | BAE31745.1 | |||
| AK159174.1 | BAE34874.1 | |||
| AK166628.1 | ||||
| AK167000.1 | BAE39179.1 | |||
| AK167129.1 | BAE39276.1 | |||
| AK167543.1 | BAE39610.1 | |||
| AK167931.1 | BAE39935.1 | |||
| AK168686.1 | BAE40532.1 | |||
| AK169166.1 | BAE40945.1 | |||
| AK170092.1 | ||||
| AK170586.1 | BAE41895.1 | |||
| CN705066.1 | ||||
| D87898.1 | BAA13490.1 |
More Information
Set Membership: |
|

Genomic Map