Assay Details
Target Gene Details
Entrez Gene ID: | 100041412 |
Gene Name: | predicted gene 11938 |
Gene Aliases: |
- |
Location: |
Chr.11:99602646-99603308 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm11938 | NM_001127354.1 | 1 | 558 | NP_001120826.1 |

Genomic Map