Assay Details
Target Gene Details
Entrez Gene ID: | 15413 |
Gene Name: | homeobox B5 |
Gene Aliases: |
AI385717, Hox-2.1 |
Location: |
Chr.11:96303512-96306121 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxb5 | NM_008268.2 | 1 | 218 | NP_032294.2 |
| AK012146.1 | BAB28059.1 | |||
| AK133512.1 | 1 | 134 | BAE21695.1 | |
| BC103595.1 | 1 | 138 | AAI03596.1 | |
| BC103596.1 | 1 | 138 | AAI03597.1 | |
| BC103604.1 | 1 | 138 | AAI03605.1 | |
| BC103607.1 | 1 | 138 | AAI03608.1 | |
| M26283.1 | 1 | 218 | AAA37842.1 |
More Information
Set Membership: |
|

Genomic Map