Assay Details
Target Gene Details
Entrez Gene ID: | 15414 |
Gene Name: | homeobox B6 |
Gene Aliases: |
Hox-2.2 |
Location: |
Chr.11:96292669-96301569 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxb6 | NM_008269.1 | 2 | 716 | NP_032295.1 |
| AK011622.1 | 2 | 816 | ||
| BC016893.1 | 2 | 716 | AAH16893.1 | |
| X56461.1 | 2 | 1224 |
More Information
Set Membership: |
|

Genomic Map