Assay Details
Target Gene Details
Entrez Gene ID: | 209586 |
Gene Name: | NudC domain containing 3 |
Gene Aliases: |
AI427847, BC024322, mKIAA1068 |
Location: |
Chr.11:6105691-6200862 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudcd3 | NM_173748.4 | 6 | 2185 | NP_776109.1 |
| AK030050.1 | 6 | 1911 | ||
| AK036694.1 | 6 | 1890 | ||
| AK049739.1 | 6 | 1897 | BAC33899.1 | |
| AK050476.1 | 5 | 1731 | BAC34277.1 | |
| AK083744.1 | 1 | 150 | ||
| AK129276.1 | 3 | 1100 | BAC98086.1 | |
| AK159391.1 | 6 | 1917 | BAE35045.1 | |
| AK159458.1 | 6 | 1917 | BAE35100.1 | |
| AK169797.1 | 6 | 1921 | BAE41373.1 | |
| AK170329.1 | 6 | 1918 | BAE41723.1 | |
| AK171224.1 | 6 | 1900 | BAE42326.1 | |
| AK181632.1 | ||||
| AK183736.1 | 1 | 286 | ||
| AK202413.1 | 1 | 283 | ||
| AK206895.1 | ||||
| AK207709.1 | 1 | 260 | ||
| AK208331.1 | 1 | 272 | ||
| AK210709.1 | ||||
| AK212632.1 | 1 | 219 | ||
| AK214979.1 | 1 | 310 | ||
| AK216990.1 | 1 | 301 | ||
| AK217536.1 | 1 | 270 | ||
| BC024322.1 | 6 | 1903 | AAH24322.3 | |
| BC037090.1 | 6 | 1884 | AAH37090.1 | |
| BC057603.1 | 5 | 1653 | AAH57603.1 |
More Information
Set Membership: |
|

Genomic Map