Assay Details
Target Gene Details
Entrez Gene ID: | 108083 |
Gene Name: | phosphatidylinositol-5-phosphate 4-kinase, type II, beta |
Gene Aliases: |
AI848124, PI5P4Kbeta, Pip5k2b, c11 |
Location: |
Chr.11:97715157-97744704 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pip4k2b | NM_054051.1 | 10 | 2532 | NP_473392.1 |
| AK089414.1 | 10 | 2521 | ||
| AK142164.1 | 1 | 1096 | ||
| AK146559.1 | 10 | 2582 | BAE27260.1 | |
| AK170392.1 | 11 | 2014 | ||
| AK171497.1 | 10 | 2525 | ||
| BC047282.1 | 10 | 2532 | AAH47282.1 |
More Information
Set Membership: |
|

Genomic Map