Assay Details
Target Gene Details
Entrez Gene ID: | 108083 |
Gene Name: | phosphatidylinositol-5-phosphate 4-kinase, type II, beta |
Gene Aliases: |
AI848124, PI5P4Kbeta, Pip5k2b, c11 |
Location: |
Chr.11:97715157-97744704 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pip4k2b | NM_054051.1 | 8 | 1081 | NP_473392.1 |
| AB054987.1 | 4 | 474 | BAB62148.1 | |
| AK089414.1 | 8 | 1069 | ||
| AK146559.1 | 8 | 1131 | BAE27260.1 | |
| AK170392.1 | 8 | 1008 | ||
| AK171497.1 | 8 | 1071 | ||
| AY050219.1 | 8 | 910 | AAL18245.1 | |
| BC047282.1 | 8 | 1081 | AAH47282.1 |
More Information
Set Membership: |
|

Genomic Map