Assay Details
Target Gene Details
Entrez Gene ID: | 72503 |
Gene Name: | RIKEN cDNA 2610507B11 gene |
Gene Aliases: |
AU022687, D11Bhm178e, D11Bhm179e, E1, Kiaa0100, mKIAA0100 |
Location: |
Chr.11:78261703-78290625 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610507B11Rik | NM_001002004.2 | 16 | 2612 | NP_001002004.2 |
| AK012059.1 | 3 | 883 | BAB28000.1 | |
| AK122206.1 | 1 | 264 | BAC65488.1 | |
| AK149719.1 | 16 | 2583 | BAE29046.1 | |
| BC040464.1 | AAH40464.1 | |||
| BC075633.1 | 16 | 2572 | AAH75633.1 | |
| X81632.1 | 1 | 521 |

Genomic Map