Assay Details
Target Gene Details
Entrez Gene ID: | 72605 |
Gene Name: | carbonic anhydrase 10 |
Gene Aliases: |
2700029L05Rik, BB085816, Ca10 |
Location: |
Chr.11:93098142-93601751 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car10 | NM_028296.3 | 9 | 3059 | NP_082572.1 |
| AK132222.1 | 9 | 2900 | BAE21042.1 | |
| CJ280653.1 | 1 | 154 |

Genomic Map