Assay Details
Target Gene Details
Entrez Gene ID: | 15525 |
Gene Name: | heat shock protein 4 |
Gene Aliases: |
70kDa, AI317151, APG-2, Hsp110, Hsp70RY, mKIAA4025 |
Location: |
Chr.11:53259814-53300479 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspa4 | NM_008300.3 | 1 | 348 | NP_032326.3 |
| AI316935.1 | 1 | 348 | ||
| AK038627.1 | 1 | 348 | ||
| AK053601.1 | 1 | 281 | ||
| AK054010.1 | 1 | 289 | BAC35620.1 | |
| AK133772.1 | 1 | 289 | ||
| AK143543.1 | 1 | 304 | ||
| AK155305.1 | 1 | 265 | BAE33178.1 | |
| AK166102.1 | 1 | 333 | ||
| AK171940.1 | 1 | 326 | ||
| AK220167.1 | 1 | 299 | BAD90352.1 | |
| BC003770.1 | 1 | 258 | AAH03770.1 | |
| D85904.1 | 1 | 50 | BAA12914.1 |

Genomic Map