Assay Details
Target Gene Details
Entrez Gene ID: | 15525 |
Gene Name: | heat shock protein 4 |
Gene Aliases: |
70kDa, AI317151, APG-2, Hsp110, Hsp70RY, mKIAA4025 |
Location: |
Chr.11:53259814-53300479 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspa4 | NM_008300.3 | 19 | 3635 | NP_032326.3 |
| AK134082.1 | 1 | 652 | BAE22005.1 | |
| AK148883.1 | 8 | 1909 | ||
| AK220167.1 | 19 | 3586 | BAD90352.1 |

Genomic Map