Assay Details
Target Gene Details
Entrez Gene ID: | 171284 |
Gene Name: | T cell immunoglobulin and mucin domain containing 2 |
Gene Aliases: |
AA816106, AU019358, AU019662, C78111, G9D3, TIM-2, Tim2 |
Location: |
Chr.11:46668960-46707104 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Timd2 | NM_001161355.1 | 3 | 404 | NP_001154827.1 |
| NM_001161356.1 | 3 | 177 | NP_001154828.1 | |
| NM_134249.5 | 3 | 286 | NP_599010.4 | |
| AB009015.1 | 3 | 177 | ||
| AF399827.1 | 3 | 138 | AAL35772.1 | |
| AF399828.1 | 3 | 138 | AAL35773.1 | |
| AK145630.1 | 3 | 165 | BAE26551.1 | |
| AK163225.1 | 3 | 165 | BAE37243.1 | |
| AK166877.1 | 3 | 158 | BAE39088.1 | |
| BC025096.1 | 3 | 216 | AAH25096.1 | |
| BC028829.1 | 3 | 248 | AAH28829.1 | |
| BY731803.1 | 3 | 286 | ||
| CB953226.1 | 3 | 407 |

Genomic Map