Assay Details
Target Gene Details
Entrez Gene ID: | 68385 |
Gene Name: | TLC domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.11:78176716-78180820 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tlcd1 | NM_001291235.1 | NP_001278164.1 | ||
| NM_001291236.1 | 1 | 344 | NP_001278165.1 | |
| NM_001291237.1 | NP_001278166.1 | |||
| NM_026708.2 | 1 | 344 | NP_080984.1 | |
| AA671983.1 | ||||
| AK002265.1 | 1 | 291 | ||
| AK002703.1 | 1 | 240 | BAB22297.1 | |
| AK046577.1 | 2 | 2224 | BAE20654.1 | |
| BC005702.1 | 1 | 236 | AAH05702.1 | |
| BC039182.1 | 1 | 316 | AAH39182.1 | |
| BG083251.2 |

Genomic Map