Assay Details
Target Gene Details
Entrez Gene ID: | 28084 |
Gene Name: | vacuolar protein sorting 25 (yeast) |
Gene Aliases: |
1110020N13Rik, AW107467, D11Wsu68e |
Location: |
Chr.11:101253611-101259547 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps25 | NM_001284411.1 | 6 | 914 | NP_001271340.1 |
| NM_001284412.1 | 6 | 1030 | NP_001271341.1 | |
| NM_026776.4 | 6 | 890 | NP_081052.2 | |
| AI849750.1 | 1 | 230 | ||
| AK002500.1 | 6 | 852 | BAB22148.1 | |
| AK003715.2 | 6 | 862 | BAB22955.1 | |
| AK005149.1 | 6 | 844 | ||
| AK082933.1 | 6 | 1009 | ||
| AK145811.1 | 6 | 861 | BAE26665.1 | |
| AK150822.1 | 6 | 872 | BAE29884.1 | |
| AK162230.1 | 6 | 1003 | ||
| AK166008.1 | 6 | 869 | BAE38516.1 | |
| AK206274.1 | 1 | 270 | ||
| AK209046.1 | 1 | 319 | ||
| AK210259.1 | 1 | 343 | ||
| AK212620.1 | ||||
| AK217046.1 | 1 | 352 | ||
| BC029181.1 | 6 | 854 | AAH29181.1 |
More Information
Set Membership: |
|

Genomic Map