Assay Details
Target Gene Details
Entrez Gene ID: | 245944 |
Gene Name: | vacuolar protein sorting 54 (yeast) |
Gene Aliases: |
Hcc8, Vps54l, mSLP8, wr |
Location: |
Chr.11:21239032-21321133 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps54 | NM_001290628.1 | 12 | 1892 | NP_001277557.1 |
| NM_139061.5 | 12 | 1928 | NP_620692.3 | |
| AF424699.2 | 12 | 1679 | AAL24808.2 | |
| AK036667.1 | 6 | 675 | ||
| AK044995.1 | 6 | 708 | BAC32176.1 | |
| AK077306.1 | 10 | 1310 | BAC36741.1 | |
| BC025012.2 | 12 | 1603 | AAH25012.1 |
More Information
Set Membership: |
|

Genomic Map