Assay Details
Target Gene Details
Entrez Gene ID: | 245944 |
Gene Name: | vacuolar protein sorting 54 (yeast) |
Gene Aliases: |
Hcc8, Vps54l, mSLP8, wr |
Location: |
Chr.11:21239032-21321133 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps54 | NM_001290628.1 | 23 | 4305 | NP_001277557.1 |
| NM_139061.5 | 23 | 4341 | NP_620692.3 | |
| AB052761.1 | 7 | 1752 | BAC53794.1 | |
| AF424699.2 | 23 | 4092 | AAL24808.2 | |
| AK036667.1 | 17 | 3088 | ||
| AK137211.1 | 9 | 1886 | BAE23271.1 | |
| AV247540.1 |
More Information
Set Membership: |
|

Genomic Map