Assay Details
Target Gene Details
Entrez Gene ID: | 276905 |
Gene Name: | armadillo repeat containing 7 |
Gene Aliases: |
A630084C13, BC023126 |
Location: |
Chr.11:115475638-115490468 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Armc7 | NM_177778.4 | 1 | 58 | NP_808446.1 |
| AK146249.1 | BAE27011.1 | |||
| BC023126.1 | 1 | 58 | AAH23126.1 |

Genomic Map